accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNHONHNCH3H3CFF

CAS RN 288150-92-5

IUPAC Name: 3-(6,8-difluoro-2-methylquinolin-4-yl)-1-[4-(dimethylamino)phenyl]urea
Synonyms: 3-(6,8-difluoro-2-methylquinolin-4-yl)-1-[4-(dimethylamino)phenyl]urea
Molecular Weight (g/mol): 356.38
Molecular Formula: C19H18F2N4O
InChi Key: JTARFZSNUAGHRB-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(NC(=O)NC2=C3C=C(F)C=C(F)C3=NC(C)=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results