accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)(R)(S)O(S)H3CHOOOHO

CAS RN 28874-45-5

IUPAC Name: (1R,2R,3S,4S)-2,3-dimethyl-7-oxabicyclo[2.2.1]heptane-2,3-dicarboxylic acid
Synonyms: cantharidic acid
Molecular Weight (g/mol): 214.22
Molecular Formula: C10H14O5
InChi Key: NMTNUQBORQILRK-YUMGAWCOSA-N
SMILES: C[C@@]1([C@H]2CC[C@H](O2)[C@@]1(C)C(O)=O)C(O)=O
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results