CAS RN 2892306-56-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ hCAII Degrader 11
Potent human carbonic anhydrase II Degrader (PROTAC™)