accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSiH3COOOCH3

CAS RN 2897-60-1

IUPAC Name: diethoxy(methyl){3-[(oxiran-2-yl)methoxy]propyl}silane
Synonyms: diethoxy(methyl)[3-(oxiran-2-ylmethoxy)propyl]silane
Molecular Weight (g/mol): 248.39
Molecular Formula: C11H24O4Si
InChi Key: OTARVPUIYXHRRB-UHFFFAOYNA-N
SMILES: CCO[Si](C)(CCCOCC1CO1)OCC
Boiling Point 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results