Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethoxy(3-glycidyloxypropyl)methylsilane, TCI America™
SDP

Catalog No. D263225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D263225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D263225ML Supplier TCI America Supplier No. D263225ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2897-60-1
Molecular Formula C11H24O4Si
Molecular Weight (g/mol) 248.39
MDL Number MFCD00046995
InChI Key OTARVPUIYXHRRB-UHFFFAOYNA-N
Synonym 3-[Diethoxy(methyl)silyl]propyl Glycidyl Ether, (3-Glycidyloxypropyl)methyldiethoxysilane
PubChem CID 62467
IUPAC Name diethoxy(methyl){3-[(oxiran-2-yl)methoxy]propyl}silane
SMILES CCO[Si](C)(CCCOCC1CO1)OCC

Specifications

Color Colorless
Boiling Point 126°C
Quantity 25 mL
Formula Weight 248.39
Physical Form Liquid
Chemical Name or Material Diethoxy(3-glycidyloxypropyl)methylsilane
EINECSNumber (2)-2072
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.