accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3HS

CAS RN 28983-37-1

IUPAC Name: 2-methyltridecane-2-thiol
Synonyms: 2-methyltridecane-2-thiol
Molecular Weight (g/mol): 230.45
Molecular Formula: C14H30S
InChi Key: LMDDHLWHSDZGIH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(C)(C)S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results