accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2902-68-3

OOIOO

CAS RN 2902-68-3

IUPAC Name: 4λ³-ioda-3,5-dioxatricyclo[5.3.1.0⁴,¹¹]undeca-1(11),7,9-triene-2,6-dione
Synonyms: 4λ³-ioda-3,5-dioxatricyclo[5.3.1.0⁴,¹¹]undeca-1(11),7,9-triene-2,6-dione
Molecular Weight (g/mol): 290.01
Molecular Formula: C8H3IO4
InChi Key: DHIWEENQJCGNME-UHFFFAOYSA-N
SMILES: O=C1O[I]2OC(=O)C3=C2C1=CC=C3
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)