accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClClClClCl

CAS RN 29082-74-4

IUPAC Name: 1,2,3,4,5-pentachloro-6-(1,2,2-trichloroethenyl)benzene
Synonyms: octachlorostyrene
Molecular Weight (g/mol): 379.69
Molecular Formula: C8Cl8
InChi Key: RUYUCCQRWINUHE-UHFFFAOYSA-N
SMILES: ClC(Cl)=C(Cl)C1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results