accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2909-38-8

ClNCO

CAS RN 2909-38-8

IUPAC Name: 1-chloro-3-isocyanatobenzene
Synonyms: 3-chlorophenyl isocyanate
Molecular Weight (g/mol): 153.57
Molecular Formula: C7H4ClNO
InChi Key: HHIRBXHEYVDUAM-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)N=C=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)