accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHO(Z)(Z)

CAS RN 29118-70-5

IUPAC Name: (5Z,9Z)-cyclododeca-5,9-diene-1,2-diol
Synonyms: (5Z,9Z)-cyclododeca-5,9-diene-1,2-diol
Molecular Weight (g/mol): 196.29
Molecular Formula: C12H20O2
InChi Key: BHYILZSOXKSYCF-GLIMQPGKNA-N
SMILES: OC1CC\C=C/CC\C=C/CCC1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results