accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2915-16-4

NNCl

CAS RN 2915-16-4

IUPAC Name: 2-chloro-4,6-diphenylpyrimidine
Synonyms: 2-chloro-4,6-diphenylpyrimidine
Molecular Weight (g/mol): 266.73
Molecular Formula: C16H11ClN2
InChi Key: QNGVEVOZKYHNGL-UHFFFAOYSA-N
SMILES: ClC1=NC(=CC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results