Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4,6-diphenylpyrimidine 98.0+%, TCI America™
SDP

Catalog No. C31235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C31235G Supplier TCI America Supplier No. C31235G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2915-16-4
Molecular Formula C16H11ClN2
Molecular Weight (g/mol) 266.728
MDL Number MFCD00030776
InChI Key QNGVEVOZKYHNGL-UHFFFAOYSA-N
PubChem CID 1804739
IUPAC Name 2-chloro-4,6-diphenylpyrimidine
SMILES C1=CC=C(C=C1)C2=CC(=NC(=N2)Cl)C3=CC=CC=C3

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 5 g
Formula Weight 266.73
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4,6-diphenylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.