CAS RN 29159-23-7
IUPAC Name:
3-aminobenzaldehyde
Synonyms:
formaldehyde, polymer with benzenamine
Molecular Weight (g/mol):
121.14
Molecular Formula:
C7H7NO
InChi Key:
SIXYIEWSUKAOEN-UHFFFAOYSA-N
SMILES:
NC1=CC=CC(C=O)=C1
1
–
1
of
1
results
1
3-Aminobenzaldehyde Polymer, TCI America™
CAS: 29159-23-7 Molecular Formula: C7H7NO Molecular Weight (g/mol): 121.139 MDL Number: MFCD00147530 InChI Key: SIXYIEWSUKAOEN-UHFFFAOYSA-N Synonym: Poly(3-aminobenzaldehyde) PubChem CID: 74366 IUPAC Name: 3-aminobenzaldehyde SMILES: C1=CC(=CC(=C1)N)C=O