accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29213-03-4

BrH2NBrNH2

CAS RN 29213-03-4

IUPAC Name: 2,6-dibromobenzene-1,4-diamine
Synonyms: 2,6-dibromobenzene-1,4-diamine
Molecular Weight (g/mol): 265.94
Molecular Formula: C6H6Br2N2
InChi Key: NIXNGYGWQMXMCA-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=C(N)C(Br)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results