accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNNHNHCl

CAS RN 2922-28-3

IUPAC Name: 7H-purin-6-amine hydrochloride
Synonyms: adenine hydrochloride
Molecular Weight (g/mol): 171.59
Molecular Formula: C5H6ClN5
InChi Key: UQVDQSWZQXDUJB-UHFFFAOYSA-N
SMILES: Cl.NC1=C2NC=NC2=NC=N1
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (5)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (5)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (5)
  • (5)