Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Adenine
GREENER_CHOICESDP

Catalog No. 420250 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-025-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 42-025-0 Supplier R&D Systems™ Supplier No. 4202/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Adenine receptor agonist

High affinity adenine receptor agonist (Ki = 18 nM at rat adenine receptor). Inhibits forskolin-stimulated cAMP formation in CHO cells transfected with the adenine receptor; also stimulates GTPγS binding (pEC50 values are 8.54 and 7.21 respectively).

Chemical Identifiers

CAS 2922-28-3
Molecular Formula C5H6ClN5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 171.59
Purity >99%
Molecular Weight (g/mol) 171.59
Chemical Name or Material Adenine
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.