accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFBrFClOF

CAS RN 292621-46-6

IUPAC Name: 3-bromo-2,4,5,6-tetrafluorobenzoyl chloride
Synonyms: 3-bromo-2,4,5,6-tetrafluorobenzoyl chloride
Molecular Weight (g/mol): 291.42
Molecular Formula: C7BrClF4O
InChi Key: FHHUFCXIDZNVQW-UHFFFAOYSA-N
SMILES: FC1=C(F)C(Br)=C(F)C(C(Cl)=O)=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results