accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 292621-58-0

IUPAC Name: 3-chloro-2,4,5,6-tetrafluorobenzoyl chloride
Synonyms: 3-chloro-2,4,5,6-tetrafluorobenzoyl chloride
Molecular Weight (g/mol): 246.97
Molecular Formula: C7Cl2F4O
InChi Key: GNXAQPALNSLOBK-UHFFFAOYSA-N
SMILES: FC1=C(F)C(Cl)=C(F)C(C(Cl)=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results