accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3ONHOH2NHNONHOOH3CCH3CH3HO

CAS RN 292632-98-5

IUPAC Name: tert-butyl N-[5-benzyl-5-({1-[(1-carbamoyl-2-phenylethyl)carbamoyl]-3-methylbutyl}carbamoyl)-3-hydroxy-1-phenylpentan-2-yl]carbamate
Synonyms: tert-butyl N-[5-benzyl-5-({1-[(1-carbamoyl-2-phenylethyl)carbamoyl]-3-methylbutyl}carbamoyl)-3-hydroxy-1-phenylpentan-2-yl]carbamate
Molecular Weight (g/mol): 672.87
Molecular Formula: C39H52N4O6
InChi Key: MURCDOXDAHPNRQ-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(=O)C(CC(O)C(CC1=CC=CC=C1)NC(=O)OC(C)(C)C)CC1=CC=CC=C1)C(=O)NC(CC1=CC=CC=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)