accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNHHN

CAS RN 294-80-4

IUPAC Name: 1,5,9-triazacyclododecane
Synonyms: 1,5,9-triazacyclododecane
Molecular Weight (g/mol): 171.29
Molecular Formula: C9H21N3
InChi Key: VQFZKDXSJZVGDA-UHFFFAOYSA-N
SMILES: C1CNCCCNCCCNC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results