accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2941-72-2

IUPAC Name: 6-methoxy-2-methyl-1,3-benzothiazole
Synonyms: 6-methoxy-2-methyl-1,3-benzothiazole
Molecular Weight (g/mol): 179.24
Molecular Formula: C9H9NOS
InChi Key: DYHLJSUORLPGNT-UHFFFAOYSA-N
SMILES: COC1=CC=C2N=C(C)SC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results