Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxy-2-methylbenzothiazole, 97%

Catalog No. AAH3242506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3242506 5 g
AAH3242503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3242506 Supplier Thermo Scientific Chemicals Supplier No. H3242506
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2941-72-2 | C9H9NOS | 179.237 g/mol

6-Methoxy-2-methylbenzothiazole was used as an inhibitor of aryl hydrocarbon hydroxylase (PB/AHH) and aminopyrine N-demethylase (ADPM) in hepatic microsomes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methoxy-2-methylbenzothiazole was used as an inhibitor of aryl hydrocarbon hydroxylase (PB/AHH) and aminopyrine N-demethylase (ADPM) in hepatic microsomes.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2941-72-2
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.237
MDL Number MFCD00005795
InChI Key DYHLJSUORLPGNT-UHFFFAOYSA-N
Synonym 6-methoxy-2-methylbenzo d thiazole, 6-methoxy-2-methylbenzothiazole, benzothiazole, 6-methoxy-2-methyl, 2-methyl-6-methoxybenzothiazole, 6-methoxy-2-methyl-benzothiazole, benzothiazole,6-methoxy-2-methyl, 6-methoxy-2-methylbenzothiazol, #, benzothiazole, 6-methoxy-2-methyl-6ci,7ci,8ci,9ci
PubChem CID 76254
IUPAC Name 6-methoxy-2-methyl-1,3-benzothiazole
SMILES CC1=NC2=C(S1)C=C(C=C2)OC

Specifications

Density 1.204
Flash Point 109°C (228°F)
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 179.24
Percent Purity 97%
Chemical Name or Material 6-Methoxy-2-methylbenzothiazole
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 220-931-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.