accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNSONHOHNN

CAS RN 294623-49-7

IUPAC Name: N-{1-[(cyanomethyl)carbamoyl]cyclohexyl}-4-[2-(4-methylpiperazin-1-yl)-1,3-thiazol-4-yl]benzamide
Synonyms: N-[1-(cyanomethylcarbamoyl)cyclohexyl]-4-[2-(4-methylpiperazin-1-yl)-1,3-thiazol-4-yl]benzamide
Molecular Weight (g/mol): 466.6
Molecular Formula: C24H30N6O2S
InChi Key: FIVYCSWOCXEWSE-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=NC(=CS1)C1=CC=C(C=C1)C(=O)NC1(CCCCC1)C(=O)NCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)