CAS RN 29509-91-9
CAS RN 29509-91-9
IUPAC Name:
4-chloro-2,6-diphenylpyrimidine
Synonyms:
4-chloro-2,6-diphenylpyrimidine
Molecular Weight (g/mol):
266.73
Molecular Formula:
C16H11ClN2
InChi Key:
MJDDVTZXYXHTRY-UHFFFAOYSA-N
SMILES:
ClC1=NC(=NC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
4-Chloro-2,6-diphenylpyrimidine 98.0+%, TCI America™

CAS: 29509-91-9 Molecular Formula: C16H11ClN2 Molecular Weight (g/mol): 266.728 MDL Number: MFCD00234890 InChI Key: MJDDVTZXYXHTRY-UHFFFAOYSA-N PubChem CID: 582041 IUPAC Name: 4-chloro-2,6-diphenylpyrimidine SMILES: C1=CC=C(C=C1)C2=CC(=NC(=N2)C3=CC=CC=C3)Cl