Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2,6-diphenylpyrimidine 98.0+%, TCI America™
SDP

Catalog No. C30165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30165G 5 g
C30161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30165G Supplier TCI America Supplier No. C30165G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29509-91-9
Molecular Formula C16H11ClN2
Molecular Weight (g/mol) 266.728
MDL Number MFCD00234890
InChI Key MJDDVTZXYXHTRY-UHFFFAOYSA-N
PubChem CID 582041
IUPAC Name 4-chloro-2,6-diphenylpyrimidine
SMILES C1=CC=C(C=C1)C2=CC(=NC(=N2)C3=CC=CC=C3)Cl

Specifications

Melting Point 106°C
Color White
Quantity 5 g
Formula Weight 266.73
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2,6-diphenylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.