CAS RN 295325-62-1
IUPAC Name:
2-amino-5-(4-methoxy-7-nitro-2,3-dihydro-1H-indol-1-yl)-5-oxopentanoic acid
Synonyms:
2-amino-5-(4-methoxy-7-nitro-2,3-dihydroindol-1-yl)-5-oxopentanoic acid
Molecular Weight (g/mol):
323.31
Molecular Formula:
C14H17N3O6
InChi Key:
GXIDBZKXGUNITQ-UHFFFAOYNA-N
SMILES:
COC1=C2CCN(C(=O)CCC(N)C(O)=O)C2=C(C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
R&D Systems™ MNI-caged-L-glutamate
CAS: 295325-62-1 Molecular Formula: C14H17N3O6