accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3SiCH3H3C

CAS RN 29681-57-0

IUPAC Name: tert-butyldimethylsilyl
Synonyms: tert-butyldimethylsilyl group
Molecular Weight (g/mol): 115.27
Molecular Formula: C6H15Si
InChi Key: ILMRJRBKQSSXGY-UHFFFAOYSA-N
SMILES: C[Si](C)C(C)(C)C
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)