Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyldimethylsilane 95.0+%, TCI America™
SDP

Catalog No. B35655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35655G Supplier TCI America Supplier No. B35655G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29681-57-0
Molecular Formula C6H15Si
Molecular Weight (g/mol) 115.271
MDL Number MFCD00010754
InChI Key ILMRJRBKQSSXGY-UHFFFAOYSA-N
Synonym tert-butyldimethylsilane, t-butyldimethylsilane, tert-butyldimethylsilyl, silane, 1,1-dimethylethyl dimethyl, t-butyldimethylsilyl, t-butyl-dimethylsilyl, t-butyl-dimethyl-silyl, tert-butyl dimethylsilane, tert-butyldimethylsilyl group
PubChem CID 9898815
IUPAC Name tert-butyl(dimethyl)silicon
SMILES CC(C)(C)[Si](C)C

Specifications

Color Colorless
Boiling Point 72°C
Quantity 5 g
UN Number 1993
Formula Weight 116.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyldimethylsilane
EINECSNumber (2)-3696
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.