Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyldimethylsilane, 95%

Catalog No. AC445710250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445710250 25 g
AC445710050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445710250 Supplier Thermo Scientific Chemicals Supplier No. 445710250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 29681-57-0 | C6H16Si | 116.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29681-57-0
Molecular Formula C6H16Si
Molecular Weight (g/mol) 116.28
InChI Key ILMRJRBKQSSXGY-UHFFFAOYSA-N
Synonym tert-butyldimethylsilane, t-butyldimethylsilane, tert-butyldimethylsilyl, silane, 1,1-dimethylethyl dimethyl, t-butyldimethylsilyl, t-butyl-dimethylsilyl, t-butyl-dimethyl-silyl, tert-butyl dimethylsilane, tert-butyldimethylsilyl group
PubChem CID 9898815
IUPAC Name tert-butyl(dimethyl)silicon
SMILES CC(C)(C)[Si](C)C

Specifications

Density 0.7010g/mL
Boiling Point 81°C to 83°C
Flash Point -11°C
Assay Percent Range 94% min. (GC)
Quantity 25 g
Specific Gravity 0.701
Formula Weight 116.28
Percent Purity 95%
Chemical Name or Material tert-Butyldimethylsilane
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.