accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2971-79-1

IUPAC Name: methyl piperidine-4-carboxylate
Synonyms: methyl piperidine-4-carboxylate
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: RZVWBASHHLFBJF-UHFFFAOYSA-N
SMILES: COC(=O)C1CCNCC1
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)