Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Piperidinecarboxylate 98.0+%, TCI America™
SDP

Catalog No. M20755G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20755G 5 g
M207525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20755G Supplier TCI America Supplier No. M20755G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2971-79-1
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00190578
InChI Key RZVWBASHHLFBJF-UHFFFAOYSA-N
Synonym methyl isonipecotate, methyl 4-piperidinecarboxylate, 4-piperidinecarboxylic acid, methyl ester, piperidine-4-carboxylic acid methyl ester, 4-piperidinecarboxylic acid methyl ester, isonipecotic acid methyl ester, methylpiperidine-4-carboxylate, methylisonipecotate, methyl piperidin-4-carboxylate, acmc-20aiv4
PubChem CID 424914
IUPAC Name methyl piperidine-4-carboxylate
SMILES COC(=O)C1CCNCC1

Specifications

Color Yellow
Boiling Point 93°C
Quantity 5 g
Formula Weight 143.19
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Methyl 4-Piperidinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.