Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl piperidine-4-carboxylate, 98%

Catalog No. AAH6175422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6175422 100 g
AAH6175414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6175422 Supplier Thermo Scientific Chemicals Supplier No. H6175422
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2971-79-1 | C7H13NO2 | 143.19 g/mol

Methyl piperidine-4-carboxylate is a reactant for C-2 arylation of piperidines through directed transition metal-catalyzed sp3 C-H activation. It is used as a reactant for synthesis of antitubercular agents, aminopyrazine inhibitors, orally available naphthyridine protein kinase D inhibitors and a scalable catalyst from asymmetric hydrogenation of sterically demanding aryl enamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl piperidine-4-carboxylate is a reactant for C-2 arylation of piperidines through directed transition metal-catalyzed sp3 C-H activation. It is used as a reactant for synthesis of antitubercular agents, aminopyrazine inhibitors, orally available naphthyridine protein kinase D inhibitors and a scalable catalyst from asymmetric hydrogenation of sterically demanding aryl enamide.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2971-79-1
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00190578
InChI Key RZVWBASHHLFBJF-UHFFFAOYSA-N
Synonym methyl isonipecotate, methyl 4-piperidinecarboxylate, 4-piperidinecarboxylic acid, methyl ester, piperidine-4-carboxylic acid methyl ester, 4-piperidinecarboxylic acid methyl ester, isonipecotic acid methyl ester, methylpiperidine-4-carboxylate, methylisonipecotate, methyl piperidin-4-carboxylate, acmc-20aiv4
PubChem CID 424914
IUPAC Name methyl piperidine-4-carboxylate
SMILES COC(=O)C1CCNCC1

Specifications

Color Colorless to Yellow
Density 1.06 g/mL
Boiling Point 85°C to 90°C
Flash Point 89°C (192°F)
Refractive Index 1.465
Quantity 100 g
Beilstein 224128
Solubility Information Slightly soluble in water.
Formula Weight 143.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl piperidine-4-carboxylate
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.