accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPSOH3CSSH3C

CAS RN 298-02-2

IUPAC Name: O,O-diethyl {[(ethylsulfanyl)methyl]sulfanyl}phosphonothioate
Synonyms: rampart timet VUAgT 182 caswell no. 660 rcra waste number P094 thimenox phorate (substance) phorat epa pesticide chemical code 057201 american cyanamid 3,911
Molecular Weight (g/mol): 260.36
Molecular Formula: C7H17O2PS3
InChi Key: BULVZWIRKLYCBC-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)SCSCC
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)