CAS RN 29925-17-5
CAS RN 29925-17-5
IUPAC Name:
4-[(3-butoxy-4-methoxyphenyl)methyl]imidazolidin-2-one
Synonyms:
4-[(3-butoxy-4-methoxyphenyl)methyl]imidazolidin-2-one
Molecular Weight (g/mol):
278.35
Molecular Formula:
C15H22N2O3
InChi Key:
PDMUULPVBYQBBK-UHFFFAOYNA-N
SMILES:
CCCCOC1=CC(CC2CNC(=O)N2)=CC=C1OC
1
–
2
of
2
results
1
Tocris Bioscience™ Ro 20-1724
CAS: 29925-17-5 Molecular Formula: C15H22N2O3
2
MilliporeSigma™ Calbiochem™ Ro-20-1724
A cell-permeable, selective inhibitor of cAMP-specific phosphodiesterase (PDE IV; IC50 = 2 μM).