accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30007-39-7

IUPAC Name: [2-(4-hydroxyphenyl)ethyl][(1-oxo-1,2,3,4-tetrahydronaphthalen-2-yl)methyl]azanium chloride
Synonyms: [2-(4-hydroxyphenyl)ethyl][(1-oxo-3,4-dihydro-2H-naphthalen-2-yl)methyl]azanium chloride
Molecular Weight (g/mol): 331.84
Molecular Formula: C19H22ClNO2
InChi Key: VCZXZECZIRGUCZ-UHFFFAOYNA-N
SMILES: [Cl-].OC1=CC=C(CC[NH2+]CC2CCC3=CC=CC=C3C2=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results