CAS RN 300576-59-4
IUPAC Name:
6-propyl-3,9-dioxa-6-azatricyclo[8.4.0.0²,⁷]tetradeca-1(10),11,13-trien-13-ol hydrochloride
Synonyms:
6-propyl-3,9-dioxa-6-azatricyclo[8.4.0.0²,⁷]tetradeca-1(10),11,13-trien-13-ol hydrochloride
Molecular Weight (g/mol):
285.77
Molecular Formula:
C14H20ClNO3
InChi Key:
DCFXOTRONMKUJB-UHFFFAOYNA-N
SMILES:
Cl.CCCN1CCOC2C1COC1=C2C=C(O)C=C1
1
–
1
of
1
results
1
R&D Systems™ (+)-PD 128907 hydrochloride
CAS: 300576-59-4 Molecular Formula: C14H20ClNO3