accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHOONONH3C

CAS RN 300816-15-3

IUPAC Name: 6-methyl-1'-[2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethyl]-1,2-dihydrospiro[3,1-benzoxazine-4,4'-piperidin]-2-one
Synonyms: 6-methyl-1'-[2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethyl]-1H-spiro[3,1-benzoxazine-4,4'-piperidin]-2-one
Molecular Weight (g/mol): 417.51
Molecular Formula: C25H27N3O3
InChi Key: ODNICNWASXKNNQ-UHFFFAOYSA-N
SMILES: CC1=C(CCN2CCC3(CC2)OC(=O)NC2=C3C=C(C)C=C2)N=C(O1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results