accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OSOOONH2

CAS RN 30203-11-3

IUPAC Name: 3-{4-[4-(3-aminophenoxy)benzenesulfonyl]phenoxy}aniline
Synonyms: 3-{4-[4-(3-aminophenoxy)benzenesulfonyl]phenoxy}aniline
Molecular Weight (g/mol): 432.49
Molecular Formula: C24H20N2O4S
InChi Key: WCXGOVYROJJXHA-UHFFFAOYSA-N
SMILES: NC1=CC(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(OC3=CC(N)=CC=C3)C=C2)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results