CAS RN 30275-80-0
CAS RN 30275-80-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Adenosine 3',5'-cyclic Monophosphate, N6-Benzoyl-, Sodium Salt
A potent cell-permeable activator of cAMP-dependent protein kinase (PKA).