accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHNOClClNH(E/Z)

CAS RN 302803-72-1

IUPAC Name: 3-(2,4-dichlorophenyl)-3-oxo-2-(4-oxo-1,2,3,4-tetrahydroquinazolin-2-ylidene)propanenitrile
Synonyms: 3-(2,4-dichlorophenyl)-3-oxo-2-(4-oxo-1,3-dihydroquinazolin-2-ylidene)propanenitrile
Molecular Weight (g/mol): 358.18
Molecular Formula: C17H9Cl2N3O2
InChi Key: SESYPWCSIZUIAS-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(C=C1)C(=O)C(C#N)=C1NC(=O)C2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results