accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3029-40-1

IUPAC Name: [(1E,3E,5E,7E)-8-phenylocta-1,3,5,7-tetraen-1-yl]benzene
Synonyms: [(1E,3E,5E,7E)-8-phenylocta-1,3,5,7-tetraen-1-yl]benzene
Molecular Weight (g/mol): 258.36
Molecular Formula: C20H18
InChi Key: ZENGMMQJMCPHTK-FPPPDJHPSA-N
SMILES: C(\C=C\C=C\C1=CC=CC=C1)=C/C=C/C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results