accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(S)(R)CH3(S)(R)CH3(R)(S)(S)(R)CH3CH3CH3(Z)

CAS RN 303-43-5

IUPAC Name: (1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl (9Z)-octadec-9-enoate
Synonyms: cholesteryl oleate cholesteryl oleate, (e)-isomer cholesteryl oleate, (e)- 5-cholesten-3b-ol 3-oleate cholesteryl [9,10-3H]oleate cholest-5-en-3b-yl cholesteryl oleate, (E)- (e)-cholesteryl oleate cholesteryl oleate, (E)-isomer cholesteroyl-oleate
Molecular Weight (g/mol): 651.12
Molecular Formula: C45H78O2
InChi Key: RJECHNNFRHZQKU-RMUVNZEASA-N
SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results