CAS RN 3030588-01-0
CAS RN 3030588-01-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ NVS-STG2
STING agonist, molecular glue that causes oligomerization of STING