accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 303134-03-4

IUPAC Name: 4-(4-methyl-1,4-diazepan-1-yl)benzoic acid hydrochloride
Synonyms: 4-(4-methyl-1,4-diazepan-1-yl)benzoic acid hydrochloride
Molecular Weight (g/mol): 270.76
Molecular Formula: C13H19ClN2O2
InChi Key: KJZVHMAKWMCIBB-UHFFFAOYSA-N
SMILES: Cl.CN1CCCN(CC1)C1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results