accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNSNNHOCH3

CAS RN 303162-79-0

IUPAC Name: N-{4-[2-ethyl-4-(3-methylphenyl)-1,3-thiazol-5-yl]pyridin-2-yl}benzamide
Synonyms: N-{4-[2-ethyl-4-(3-methylphenyl)-1,3-thiazol-5-yl]pyridin-2-yl}benzamide
Molecular Weight (g/mol): 399.51
Molecular Formula: C24H21N3OS
InChi Key: HEKAIDKUDLCBRU-UHFFFAOYSA-N
SMILES: CCC1=NC(=C(S1)C1=CC(NC(=O)C2=CC=CC=C2)=NC=C1)C1=CC(C)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)