accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONCH3OONa

CAS RN 30377-07-2

IUPAC Name: sodium 2-(N-methyldecanamido)acetate
Synonyms: sodium 2-(N-methyldecanamido)acetate
Molecular Weight (g/mol): 265.33
Molecular Formula: C13H24NNaO3
InChi Key: LXXIWMVZOWQLIH-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCC(=O)N(C)CC([O-])=O
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results