accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrHNONOOCl

CAS RN 304672-88-6

IUPAC Name: N-(3-bromophenyl)-2-chloro-4-nitrobenzamide
Synonyms: N-(3-bromophenyl)-2-chloro-4-nitrobenzamide
Molecular Weight (g/mol): 355.57
Molecular Formula: C13H8BrClN2O3
InChi Key: IGFNILVZYOMTAJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(C=C1)C(=O)NC1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results