accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHClClFNNF(E)

CAS RN 30484-77-6

IUPAC Name: dihydrogen 1-[bis(4-fluorophenyl)methyl]-4-[(2E)-3-phenylprop-2-en-1-yl]piperazine dichloride
Synonyms: flunarizine
Molecular Weight (g/mol): 477.42
Molecular Formula: C26H28Cl2F2N2
InChi Key: RXKMOPXNWTYEHI-RDRKJGRWSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].FC1=CC=C(C=C1)C(N1CCN(C\C=C\C2=CC=CC=C2)CC1)C1=CC=C(F)C=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Product Line 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results