Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flunarizine dihydrochloride, R&D Systems™
SDP

Catalog No. 0522500 Shop All Novus Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-225-00 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 05-225-00 Supplier R&D Systems Supplier No. 0522/500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Dual Na+/Ca2+ channel (T-type) blocker

Dual Na+/Ca2+ channel blocker; a cerebral and peripheral vasodilator. Neuroprotective.

Chemical Identifiers

CAS 30484-77-6
Molecular Formula C26H28Cl2F2N2
Molecular Weight (g/mol) 477.42
MDL Number MFCD00058198
InChI Key RXKMOPXNWTYEHI-RDRKJGRWSA-N
Synonym flunarizine dihydrochloride, flunarizine hydrochloride, flunarizine 2hcl, flunarizine hcl, flunarizine hydrochloride usan:jan, 1-bis 4-fluorophenyl methyl-4-cinnamylpiperazine dihydrochloride, e-1-bis-p-fluorophenyl methyl-4-cinnamylpiperazine dihydrochloride, e-1-bis 4-fluorophenyl methyl-4-3-phenyl-2-propenyl piperazine dihydrochloride, e-1-bis 4-fluorophenyl methyl-4-cinnamylpiperazine dihydrochloride
PubChem CID 5282407
IUPAC Name dihydrogen 1-[bis(4-fluorophenyl)methyl]-4-[(2E)-3-phenylprop-2-en-1-yl]piperazine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].FC1=CC=C(C=C1)C(N1CCN(C\C=C\C2=CC=CC=C2)CC1)C1=CC=C(F)C=C1

Specifications

Quantity 500 mg
Formula Weight 477.42
Percent Purity >98%
Product Line Tocris
Chemical Name or Material Flunarizine dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.