Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flunarizine dihydrochloride, 99%

Catalog No. AAJ6296903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6296903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6296903 Supplier Thermo Scientific Chemicals Supplier No. J6296903
  • $105.40
    $83.65 / Each of 1

    Save $21.75

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 30484-77-6 | C26H28Cl2F2N2 | 477.42 g/mol

A dual sodium/calcium channel blocker; a cerebral and peripheral vasodilator

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30484-77-6
Molecular Formula C26H28Cl2F2N2
Molecular Weight (g/mol) 477.42
MDL Number MFCD00058198
InChI Key RXKMOPXNWTYEHI-RDRKJGRWSA-N
Synonym flunarizine dihydrochloride, flunarizine hydrochloride, flunarizine 2hcl, flunarizine hcl, flunarizine hydrochloride usan:jan, 1-bis 4-fluorophenyl methyl-4-cinnamylpiperazine dihydrochloride, e-1-bis-p-fluorophenyl methyl-4-cinnamylpiperazine dihydrochloride, e-1-bis 4-fluorophenyl methyl-4-3-phenyl-2-propenyl piperazine dihydrochloride, e-1-bis 4-fluorophenyl methyl-4-cinnamylpiperazine dihydrochloride
PubChem CID 5282407
IUPAC Name dihydrogen 1-[bis(4-fluorophenyl)methyl]-4-[(2E)-3-phenylprop-2-en-1-yl]piperazine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].FC1=CC=C(C=C1)C(N1CCN(C\C=C\C2=CC=CC=C2)CC1)C1=CC=C(F)C=C1

Specifications

Color White
Quantity 1 g
Beilstein 4284243
Merck Index 14,4144
Formula Weight 477.4
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Flunarizine dihydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 250-216-6
RTECSNumber TK9187000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.